Merck
  • Small molecule inhibition of a Group II chaperonin: pinpointing a loop region within the equatorial domain as necessary for protein refolding.
Small molecule inhibition of a Group II chaperonin: pinpointing a loop region within the equatorial domain as necessary for protein refolding.
Archives of biochemistry and biophysics (2008-11-04)
Lisa M Bergeron, David L Shis, Lizabeth Gomez, Douglas S Clark
ZUSAMMENFASSUNG
The functionality of regions within the equatorial domain of Group II chaperonins is poorly understood. Previously we showed that a 70 amino acid sequence within this domain on the single-subunit recombinant thermosome from Methanocaldococcus jannaschii (rTHS) contains residues directly responsible for refolding protein substrates [L.M. Bergeron, C. Lee, D.S. Clark, Identification of a critical chaperoning region on an archaeal recombinant thermosome, Biochem. Biophys. Res. Commun. 369 (2008) 707-711]. In the present study, 6-aminopenicillanic acid (6-APA) was found to bind to rTHS and inhibit it from refolding proteins. Fluorescence anisotropy was used to measure a 6-APA/rTHS dissociation constant of 17.1 microM and verify that the binding site is within the first 70 amino-terminal rTHS residues. Docking simulations point to a specific loop region at residues 53-57 on rTHS as the most likely binding region. This loop region is located within the oligomeric association sites of the wild-type thermosome. These results implicate a specific equatorial region of Group II chaperonins in the refolding of proteins, and suggest its importance in conformational changes that accompany chaperone function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(+)-6-Aminopenicillansäure, 96%