Merck
  • Ten novel HMGCL mutations in 24 patients of different origin with 3-hydroxy-3-methyl-glutaric aciduria.
Ten novel HMGCL mutations in 24 patients of different origin with 3-hydroxy-3-methyl-glutaric aciduria.
Human mutation (2009-01-30)
Sebastián Menao, Eduardo López-Viñas, Cecilia Mir, Beatriz Puisac, Esther Gratacós, María Arnedo, Patricia Carrasco, Susana Moreno, Mónica Ramos, María Concepción Gil, Angeles Pié, Antonia Ribes, Celia Pérez-Cerda, Magdalena Ugarte, Peter T Clayton, Stanley H Korman, Dolors Serra, Guillermina Asins, Feliciano J Ramos, Paulino Gómez-Puertas, Fausto G Hegardt, Nuria Casals, Juan Pié
ZUSAMMENFASSUNG
3-Hydroxy-3-methylglutaric aciduria is a rare autosomal recessive genetic disorder that affects ketogenesis and L-leucine catabolism. The clinical acute symptoms include vomiting, convulsions, metabolic acidosis, hypoketotic hypoglycaemia and lethargy. To date, 33 mutations in 100 patients have been reported in the HMGCL gene. In this study 10 new mutations in 24 patients are described. They include: 5 missense mutations: c.109G>A, c.425C>T, c.521G>A, c.575T>C and c.598A>T, 2 nonsense mutations: c.242G>A and c.559G>T, one small deletion: c.853delC, and 2 mutations in intron regions: c.497+4A>G and c.750+1G>A. Two prevalent mutations were detected, 109G>T (E37X) in 38% of disease alleles analyzed and c.504_505delCT in 10% of them. Although patients are mainly of European origin (71%) and mostly Spanish (54%), the group is ethnically diverse and includes, for the first time, patients from Pakistan, Palestine and Ecuador. We also present a simple, efficient method to express the enzyme and we analyze the possible functional effects of missense mutations. The finding that all identified missense mutations cause a >95% decrease in the enzyme activity, indicates that the disease appears only in very severe genotypes."
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Hydroxy-3-methyl-glutarsäure, ≥95%