Merck
Molecular recognition of pyridine N-oxides in water using calix[4]pyrrole receptors.
Journal of the American Chemical Society (2009-02-17)
Begoña Verdejo, Guzmán Gil-Ramírez, Pablo Ballester
ZUSAMMENFASSUNG
The incorporation of four carboxylic acids or four amino groups to the upper rim of an aryl extended calix[4]pyrrole produces water-soluble receptors which are able to effectively bind aromatic N-oxides in water by a combination of hydrogen bonding, pi-pi, CH-pi, and hydrophobic interactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin-N-oxid, 95%