Merck
[Studies on HPLC fingerprint chromatogram of Folium Fici Microcarpa].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2009-02-24)
Zhi-Jian Fang, Zhen Dai, Shu-Yuan Li
ZUSAMMENFASSUNG
To establish a sensitive and specific method for quality control of Folium Fici Microcarpa, HPLC method was applied for studies on the fingerprint chromatogram of Folium Fici Microcarpa. Isovitexin was used as reference substance to evaluate the chromatogram of 10 samples from different regions and 12 samples collected in different months. The result revealed that all the chromatographic peaks were seperated efficiently. There were 17 common peaks showed in the fingerprint chromatogram. The method of fingerprint chromatogram with characteristic and specificity will be used to identify the quality and evaluate different origins and collection period of Folium Fici Microcarpa.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isovitexin, ≥98.0% (HPLC)
Isovitexin, primary reference standard