Merck
  • Alternative vs. conventional treatment given on-demand for gastroesophageal reflux disease: a randomised controlled trial.
Alternative vs. conventional treatment given on-demand for gastroesophageal reflux disease: a randomised controlled trial.
BMC complementary and alternative medicine (2009-02-25)
Per G Farup, Mathis Heibert, Victor Høeg
ZUSAMMENFASSUNG
Alternative treatments are commonly used for various disorders and often taken on-demand. On-demand treatment of gastroesophageal reflux disease (GERD) with pharmaceutical products is an established, cost-effective strategy. Comparisons between alternative medicine and pharmaceutical products are rare. The aim of this trial was to compare on-demand treatment with a pectin-based, raft-forming, natural, anti-reflux agent (PRA) with that of esomeprazole 20 mg (Eso20) in patients with mild/moderate GERD. Patients with mild/moderate GERD were randomised to a six weeks' on-demand treatment with PRA or Eso20 in a pragmatic, open, multicentre trial. Overall satisfaction with treatment, satisfactory relief on a weekly basis, reflux symptoms, and treatment preferences were noted. Seventy-seven patients were included in the analyses. Eso20 was significantly superior to PRA for proportion of overall satisfied patients (92% and 58% respectively; p = 0.001), reduction of symptoms (mean symptom scores at the end 5.9 and 8.0 respectively; p = 0.019), proportion of weeks of satisfactory relief (89% and 62% respectively; p = 0.008) and proportion preferring continuation with the same treatment (85% and 42% respectively; p < 0.001). Older patients were more satisfied than younger, and patients preferring on-demand treatment had lower symptom scores at inclusion than those preferring regular treatment. On-demand treatment with esomeprazole 20 mg was clearly superior to the pectin-based raft-forming agent. Most patients preferred on-demand treatment to regular treatment. Those preferring regular therapy had significantly more symptoms at inclusion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumbicarbonat, ACS reagent, 99.7%, powder, crystals or granules
Sigma-Aldrich
Kaliumbicarbonat, ≥99.95% trace metals basis, 99.7-100.5% dry basis
Sigma-Aldrich
Kaliumbicarbonat, BioUltra, ≥99.5% (T)