Merck
  • Effects of solvents and salt on the thermal stability of lithiated graphite used in lithium ion battery.
Effects of solvents and salt on the thermal stability of lithiated graphite used in lithium ion battery.
Journal of hazardous materials (2009-03-06)
Qingsong Wang, Jinhua Sun, Chunhua Chen
ZUSAMMENFASSUNG
The thermal stability of lithiated graphite in the presence of solvents, electrolytes and LiPF(6) salt was studied using C80 micro-calorimeter. The presence of cyclic carbonates or linear carbonates increases the activity of Li(x)C(6)-solvent coexisting system, especially for the Li(x)C(6)-linear carbonates one. LiPF(6) was detected that it increases the activity greatly of its coexisting system with lithiated graphite. The coexisting system of Li(x)C(6) with the electrolyte of LiPF(6)/ethylene carbonate+diethyl carbonate shows less thermal stability, which is attributed to the activity between diethyl carbonate and Li(x)C(6). This also agrees with the experiment result of Li(x)C(6)-diethyl carbonate coexisting system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylencarbonat, 98%
Sigma-Aldrich
Diethylcarbonat, 99%
Sigma-Aldrich
Diethylcarbonat, anhydrous, ≥99%