Merck
Evaluation of the peroxynitrite scavenging activity of heat-processed ginseng.
Journal of medicinal food (2009-03-21)
K S Kang, T Tanaka, E J Cho, T Yokozawa
ZUSAMMENFASSUNG
To ascertain the principal active peroxynitrite (ONOO(-)) scavenging components of heat-processed Panax ginseng C.A. Meyer (sun ginseng [SG]), the ONOO(-) scavenging activities of fractions and components of SG were compared. The results demonstrated that the ONOO(-) scavenging ability of SG was due to its ether fraction containing phenolic compounds. High-performance liquid chromatography analysis and ONOO(-) scavenging activity tests of the phenolic acids contained in SG identified vanillic acid, ferulic acid, p-coumaric acid, syringic acid, and maltol as the main active ONOO(-) scavenging components of SG. The ONOO(-) scavenging activities of phenolic acids and maltol were dependent on the degrees of their proton donating ability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyron, 99%
Sigma-Aldrich
Maltol, natural, FG