Merck
  • A simplified green chemistry approaches to synthesis of 2-substituted 1,2,3-triazoles and 4-amino-5-cyanopyrazole derivatives conventional heating versus microwave and ultrasound as ecofriendly energy sources.
A simplified green chemistry approaches to synthesis of 2-substituted 1,2,3-triazoles and 4-amino-5-cyanopyrazole derivatives conventional heating versus microwave and ultrasound as ecofriendly energy sources.
Ultrasonics sonochemistry (2009-04-07)
Khadijah M Al-Zaydi
ZUSAMMENFASSUNG
Cyanoacetamides 3 were prepared via reacting ethyl cyanoacetate with benzylamine. Yields and reaction times needed for reaction completion at room temperature, by microwaves (muomega) heating and under ultrasound (US) irradiations are compared. The formed cyanoacetamides were coupled with aromatic diazonium salts and the formed arylhydrazones were used as precursors to title triazoles and pyrazoles via reacting the former with hydroxylamine and chloroacetonitrile. Yields of products formed via conventional heating are compared with those of muomega and US irradiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyanoacetamid, 99%