- Ocular injury with high-pressure paint: a case report.
Ocular injury with high-pressure paint: a case report.
Archives of environmental & occupational health (2009-04-28)
Chitrabhanu Bandyopadhyay, Arjit Mitra, Rosalind J Harrison
PMID19395345
ZUSAMMENFASSUNG
Chemical injury to the eye is an ophthalmic emergency that can be caused by a work-related accident or common household products. Uradil SZ261 G3Z-65 is a resin containing butyl gycol (2-butoxyethanol) and ethyl ethanol amine (2-dimethyaminoethanol). It is used in stoving enamels for interior and exterior use. The authors report a case of ocular surface injury with a highpressure industrial spray paint-Waterborne Stoving Gentian Blue containing Uradil.