Merck
  • Comparative NMR studies of diffusional water permeability of red blood cells from different species: XV. Agile wallaby (Macropus agilis), red-necked wallaby (Macropus rufogriseus) and Goodfellow's tree kangaroo (Dendrolagus goodfellowi).
Comparative NMR studies of diffusional water permeability of red blood cells from different species: XV. Agile wallaby (Macropus agilis), red-necked wallaby (Macropus rufogriseus) and Goodfellow's tree kangaroo (Dendrolagus goodfellowi).
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2009-05-26)
Gheorghe Benga, Bogdan E Chapman, Philip W Kuchel
ZUSAMMENFASSUNG
The water diffusional permeability (P(d)) of red blood cells (RBC) from agile wallaby (Macropus agilis), red-necked wallaby (Macropus rufogriseus) and Goodfellow's tree kangaroo (Dendrolagus goodfellowi) was monitored using an Mn(2+)-doping (1)H nuclear magnetic resonance (NMR) technique at 400 MHz. The P(d) (cm s(-1)) values of agile wallaby RBCs were 7.5 x 10(-3) at 25 degrees C, 9 x 10(-3) at 30 degrees C, 11 x 10(-3) at 37 degrees C, and 13 x 10(-3) at 42 degrees C. The inhibitory effect of a mercury-containing sulfhydryl (SH)-modifying reagent p-chloromercuribenzoate (PCMB) on agile wallaby RBCs was investigated. The maximal inhibition was reached in 90 min at 37 degrees C with 2 mmol L(-1) PCMB. The value of maximal inhibition was approximately 63% when measured at 25 degrees C, approximately 52% at 37 degrees C and approximately 45% at 42 degrees C. The lowest value of P(d) (corresponding to the basal permeability to water) was approximately 3 x 10(-3) cm s(-1) at 25 degrees C. For the RBCs from red-necked wallaby (M. rufogriseus) the values of P(d) (cm s(-1)) were 7 x 10(-3) at 25 degrees C, 8 x 10(-3) at 30 degrees C, 10 x 10(-3) at 37 degrees C, and 12 x 10(-3) at 42 degrees C. Higher values of P(d) (cm s(-1)) were found for the RBCs from Goodfellow's tree kangaroo (D. goodfellowi): 8.5 x 10(-3) at 25 degrees C, 10 x 10(-3) at 30 degrees C, 13 x 10(-3) at 37 degrees C, and 15 x 10(-3) at 42 degrees C. The mean values of the activation energy of water diffusion (E(a,d)) were approximately 25 kJ mol(-1) for RBCs from the agile wallaby and tree kangaroo, respectively, and approximately 23 kJ mol(-1) for RBCs from red-necked wallaby. The values of E(a,d) increased after exposure of agile wallaby RBCs to PCMB, reaching a value of approximately 43-46 kJ mol(-1) when the maximal inhibition of P(d) was achieved.