Merck
  • Preparation, in vitro and in vivo evaluation of solid-state self-nanoemulsifying drug delivery system (SNEDDS) of vitamin A acetate.
Preparation, in vitro and in vivo evaluation of solid-state self-nanoemulsifying drug delivery system (SNEDDS) of vitamin A acetate.
Journal of drug targeting (2009-06-02)
Ehab I Taha, Saleh A Al-Suwayeh, Md Khalid Anwer
ZUSAMMENFASSUNG
Vitamin A self-nanoemulsifying drug delivery system (SNEDDS), which comprises soybean oil, Cremophor EL, and Capmul MCM-C8, was prepared and mixed with different grades of Avicel to produce homogenized powders. The resultant powders were compressed into tablets. The prepared tablets were characterized for their thickness, hardness, friability, disintegration time, and dissolution rate. In addition, the relative bioavailability of the tablets in comparison to solid-state Vitamin A oily solution (SSVAOS) tablets was investigated in rats. Vitamin A dissolution rate was markedly different from one formulation to another. From the bioavailability data, it was observed that Vitamin A SNEDD tablets have higher bioavailability (relative bioavailability 143.68%) compared with SSVAOS tablets. The AUC and C(max) of Vitamin A SNEDD tablets were found to be significantly different from that of SSVAOS tablets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Retinylacetat (Vitamin-A-Acetat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinylacetat, synthetic, crystalline solid or supercooled liquid
Supelco
Retinylacetat, analytical standard
Sigma-Aldrich
Retinylacetat, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinylacetat, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Retinolacetat, European Pharmacopoeia (EP) Reference Standard