Merck
  • Enhancing thermostability of a Rhizomucor miehei lipase by engineering a disulfide bond and displaying on the yeast cell surface.
Enhancing thermostability of a Rhizomucor miehei lipase by engineering a disulfide bond and displaying on the yeast cell surface.
Applied microbiology and biotechnology (2009-06-18)
Zhen-lin Han, Shuang-yan Han, Sui-ping Zheng, Ying Lin
ZUSAMMENFASSUNG
To increase the thermostability of Rhizomucor miehei lipase, the software Disulfide by Design was used to engineer a novel disulfide bond between residues 96 and 106, and the corresponding double cysteine mutants were constructed. The R. miehei lipase mutant could be expressed by Pichia pastoris in a free secreted form or could be displayed on the cell surface. The new disulfide bond spontaneously formed in the mutant R. miehei lipase. Thermostability was examined by measuring of hydrolysis activity using 4-nitrophenyl caprylate as a substrate. The engineered disulfide bond contributed to thermostability in the free form of the R. miehei lipase variant. The variant displayed on the yeast cell surface had significantly increased residual hydrolytic activity in aqueous solution after incubation at 60 degrees C for 5 h and increased synthetic activity in organic solvent at 60 degrees C. These results indicated that yeast surface display might improve the stability of R. miehei lipase, as well as amplifying the thermostability through the engineered disulfide bond.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caprylsäure-4-nitrophenylester, ≥90.0% (GC)