Merck
  • Effect of Nilzan and albendazole on the absorptive surfaces of Haemonchus contortus (Nematoda)--a histoenzymic study.
Effect of Nilzan and albendazole on the absorptive surfaces of Haemonchus contortus (Nematoda)--a histoenzymic study.
Angewandte Parasitologie (1990-08-01)
M Kaur, M L Sood
ZUSAMMENFASSUNG
Haemonchus contortus, incubated in 10 micrograms/ml and 50 micrograms/ml concentrations of Nilzan and albendazole in Tyrode solution were stained for histoenzymatic demonstration of various phosphatases, oxido-reductases and esterases. The intestine showed major alterations after drug treatments. The alkaline phosphatases (AkPase), adenosine triphosphatase (ATPase), glucose-6-phosphatase, succinic dehydrogenase (SDH), glutamate dehydrogenase (GDH), reduced nicotinamide adenine dinucleotide phosphate diaphorase and reduced nicotinamide adenine dinucleotide diaphorase showed a decreased activity in intestine after Nilzan treatment, whereas lactic dehydrogenase (LDH), glucose-6-phosphate dehydrogenase (G-6-PD) and monoamine oxidase resisted increased reaction. The albendazole treatment resulted in altered distribution pattern of the AkPase, ATPase, SDH, and GDH; while LDH, G-6-PD, and non-specific esterases exhibited slightly enhanced activity in the epithelium. The functional significance of these changes has been fully discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Oxyclozanid, VETRANAL®, analytical standard