Merck
  • Triflusal and aspirin have different effects on inflammatory biomarkers measured in patients with acute ischemic stroke.
Triflusal and aspirin have different effects on inflammatory biomarkers measured in patients with acute ischemic stroke.
Cerebrovascular diseases (Basel, Switzerland) (2009-07-31)
José Alvarez-Sabín, Anna Penalba, Manolo Quintana, Jerzy Krupinski, Lidia García-Bonilla, Joan Montaner
ZUSAMMENFASSUNG
Triflusal is a 4-fluoromethyl derivative of salicylic acid used for secondary prevention of ischemic stroke. Recent experimental data (permanent middle cerebral artery occlusion in rats) have shown a possible role of triflusal in neuroprotection through inhibition of inflammatory pathways. To explore whether triflusal may modulate those pathways in human stroke, evolution of several inflammation markers (pro-inflammatory, adhesion molecules, chemokines, metalloproteinases, apoptosis and angiogenesis-related biomarkers) and neurological outcome were evaluated at baseline, and at days 1, 3, 7 and 90 in a pilot study in which 30 patients with acute ischemic stroke were randomly allocated to receive triflusal or aspirin. An increase in IL-6 level was found in the aspirin group when compared to the triflusal group at the third and seventh day (p < 0.05). FGF-basic level was significantly increased at days 1 and 90 in the triflusal group (p = 0.040). The triflusal group also had higher levels of MIP-1alpha and MIP-1beta (p < 0.05) at day 1. Also among triflusal-treated patients, MCP-1 and TARC levels were increased as compared with the aspirin group at day 90 (p < 0.05). Interestingly, some of those markers modified by triflusal (MCP-1 and IL-6) were associated with neurological outcome: higher MCP-1 measured at day 3 among patients who improve at day 7 [462.3 (419.2-735.2) vs. 285 (242.1-428.2), p < 0.05], and lower levels of IL-6 at day 3 among patients who improve at day 90 [24.8 (5.6-77.3) vs. 5.4 (2.0-13.8), p < 0.05]. Triflusal modulates additional mechanisms to those of aspirin [pro-inflammatory (IL-6) and chemokine (MIP-1 and MCP-1) pathways] that could participate in the ischemic damage process following human acute stroke.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triflusal, ≥98% (HPLC), powder