Merck
Zn-mediated electrochemical allylation of aldehydes in aqueous ammonia.
Chemical communications (Cambridge, England) (2009-08-08)
Jing-Mei Huang, Yi Dong
ZUSAMMENFASSUNG
An efficient electrosynthesis of homoallylic alcohols from allylic bromides and aldehydes in aqueous ammonia is achieved in an undivided cell fitted with a pair of zinc electrodes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allylbromid, reagent grade, 97%, contains ≤1000 ppm propylene oxide as stabilizer
Sigma-Aldrich
Allylbromid, ReagentPlus®, 99%, contains ≤1000 ppm propylene oxide as stabilizer