Merck
  • The utilization of gum tragacanth to improve the growth of Rhodotorula aurantiaca and the production of gamma-decalactone in large scale.
The utilization of gum tragacanth to improve the growth of Rhodotorula aurantiaca and the production of gamma-decalactone in large scale.
Applied biochemistry and biotechnology (2009-08-15)
Mohamed Alchihab, Jacqueline Destain, Mario Aguedo, Jean-Paul Wathelet, Philippe Thonart
ZUSAMMENFASSUNG
The production of gamma-decalactone and 4-hydroxydecanoic acid by the psychrophilic yeast R. aurantiaca was studied. The effect of both compounds on the growth of R. aurantiaca was also investigated and our results show that gamma-decalactone must be one of the limiting factors for its production. The addition of gum tragacanth to the medium at concentrations of 3 and 4 g/l seems to be an adequate strategy to enhance gamma-decalactone production and to reduce its toxicity towards the cell. The production of gamma-decalactone and 4-hydroxydecanoic acid was significantly higher in 20-l bioreactor than in 100-l bioreactor. By using 20 g/l of castor oil, 6.5 and 4.5 g/l of gamma-decalactone were extracted after acidification at pH 2.0 and distillation at 100 degrees C for 45 min in 20- and 100-l bioreactors, respectively. We propose a process at industrial scale using a psychrophilic yeast to produce naturally gamma-decalactone from castor oil which acts also as a detoxifying agent; moreover the process was improved by adding a natural gum.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tragant
Sigma-Aldrich
γ-Decalacton, ≥98%, FG, FCC
Sigma-Aldrich
γ-Decanolacton, 98%