Merck
  • 4-Amino-1,8-naphthalimide-based Tröger's bases as high affinity DNA targeting fluorescent supramolecular scaffolds.
4-Amino-1,8-naphthalimide-based Tröger's bases as high affinity DNA targeting fluorescent supramolecular scaffolds.
Organic letters (2009-08-18)
Emma B Veale, Daniel O Frimannsson, Mark Lawler, Thorfinnur Gunnlaugsson
ZUSAMMENFASSUNG
The synthesis and photophysical and biological investigation of fluorescent 1,8-naphthalimide conjugated Troger's bases 1-3 are described. These structures bind strongly to DNA in competitive media at pH 7.4, with concomitant modulation in their fluorescence emission. These structures also undergo rapid cellular uptake, being localized within the nucleus within a few hours, and are cytotoxic against HL60 and (chronic myeloid leukemia) K562 cell lines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(2-Aminoethyl)-morpholin, 99%
Sigma-Aldrich
4-Amino-1,8-Naphthalimid