- Structural studies of surfactants at the oil-water interface by neutron reflectometery.
Structural studies of surfactants at the oil-water interface by neutron reflectometery.
Langmuir : the ACS journal of surfaces and colloids (2009-08-29)
A Zarbakhsh, J R P Webster, J Eames
PMID19714822
ZUSAMMENFASSUNG
We report information regarding the structure at the interface between hexadecane and an aqueous solution of trimethyl tetradecyl ammonium bromide (C14TAB) with appropriate deuterium labeling. We also report the role of the headgroup size and the charge on structures at the oil-water interface by comparing the data for C14TAB to that for a deuterated trimethyl tetradecyl ammonium sulfate (C14TAS) solution at the oil-water interface. As the charge on the counterion increases, this results in a reduction in the limiting area per molecule and an increase in the adsorbed amount at the oil-water interface.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Myristyltrimethylammoniumbromid, suitable for ion pair chromatography, LiChropur™, ≥99.0% (AT)