- Screening for reactive metabolites using electro-optical arrays featuring human liver cytosol and microsomal enzyme sources and DNA.
Screening for reactive metabolites using electro-optical arrays featuring human liver cytosol and microsomal enzyme sources and DNA.
Chemical communications (Cambridge, England) (2009-09-03)
Linlin Zhao, John B Schenkman, James F Rusling
PMID19724793
ZUSAMMENFASSUNG
We demonstrate for the first time the combination of human liver cytosol and microsomal enzyme sources into an electro-optical array to screen for reactive metabolites produced in multi-enzyme metabolic processes.
MATERIALIEN