Merck
  • Stereoselective disposition of carvedilol in man after intravenous and oral administration of the racemic compound.
Stereoselective disposition of carvedilol in man after intravenous and oral administration of the racemic compound.
European journal of clinical pharmacology (1990-01-01)
G Neugebauer, W Akpan, B Kaufmann, K Reiff
ZUSAMMENFASSUNG
The racemic compound carvedilol is a multiple-action oral antihypertensive drug that exhibits both vasodilator and non-selective beta-adrenergic blocking activities. The effects of the levorotatory S-enantiomer [S(-)-CARV] are vasodilatation and beta-blockade. The R(+)-enantiomer [R(+)-CARV] is a pure vasodilating agent. Quantitative determination of the enantiomers in human plasma by HPLC was carried out after formation of diastereoisomers with the chiral reagent 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate (GITC). The pharmacokinetics of the enantiomers were studied following i.v. (12.5 mg in 1 h) and p.o. (50 mg) administration of racemic carvedilol in ten healthy male subjects according to a randomized crossover design. The AUCs of S(-)-CARV were significantly lower than those of R(+)-CARV after both i.v. and p.o. administration. The systemic clearance of the two enantiomers was significantly different, whereas half-lives and apparent distribution volumes were comparable. Following p.o. administration, the absolute bioavailability (31.1% and 15.1%, respectively) and maximal plasma concentrations of R(+)-CARV were twice those of S(-)-CARV. A similar difference was found in the half-lives. A close correlation existed between enantiomeric ratios after i.v. and after p.o. administration, demonstrating slight intraindividual variability. The preferential systemic clearance of the S(-)-enantiomer suggests stereoselective hepatic metabolism of carvedilol, becoming especially apparent after p.o. administration. The small intrasubject variability in enantiomer ratios indicates a relatively constant relation of beta-blockade to vasodilation during chronic treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl-isothiocyanat, derivatization grade (chiral), LiChropur, ≥98.0% (HPLC)