Merck
  • N-Nitrosopiperidine and N-Nitrosodibutylamine induce apoptosis in HepG2 cells via the caspase dependent pathway.
N-Nitrosopiperidine and N-Nitrosodibutylamine induce apoptosis in HepG2 cells via the caspase dependent pathway.
Cell biology international (2009-09-15)
Almudena García, Paloma Morales, Joseph Rafter, Ana I Haza
ZUSAMMENFASSUNG
The human hepatoma cell line (HepG2) exhibited a dose and time-dependent apoptotic response following treatment with N-Nitrosopiperidine (NPIP) and N-Nitrosodibutylamine (NDBA), two recognized human carcinogens. Our results showed a significant apoptotic cell death (95%) after 24h treatment with NDBA (3.5 mM), whereas it was necessary to use high doses of NPIP (45 mM) to obtain a similar percentage of apoptotic cells (86%). In addition, both extrinsic (caspase-8) and intrinsic pathway (caspase-9) could be implicated in the N-Nitrosamines-induced apoptosis. This study also addresses the role of reactive oxygen species (ROS) as intermediates for apoptosis signaling. A significant increase in ROS levels was observed after NPIP treatment, whereas NDBA did not induce ROS. However, N-acetylcysteine (NAC) did not block NPIP-induced apoptosis. All these findings suggest that NPIP and NDBA induce apoptosis in HepG2 cells via a pathway that involves caspases but not ROS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
1-Nitrosopiperidin -Lösung, certified reference material, 5000 μg/mL in methanol