Merck
  • [Determination of optical purity of alpha-phenylethylamine by high performance liquid chromatography with pre-column derivatization].
[Determination of optical purity of alpha-phenylethylamine by high performance liquid chromatography with pre-column derivatization].
Se pu = Chinese journal of chromatography (2009-10-07)
Jinzhao Wang, Su Zeng, Danhua Wang, Gongyun Hu
ZUSAMMENFASSUNG
A simple pre-column derivatization-high performance liquid chromatographic (HPLC) method was established for the determination of optical purity of alpha-phenylethylamine. The enantiomers of alpha-phenylethylamine were derivatized with 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate (GITC). The resulted diastereoisomers were separated on an Agilent Zorbax C18 column (250 mm x 4.6 mm, 5 microm) with a mobile phase of methanol-phosphate buffer (1.36 g/L aqueous solution of potassium dihydrogen phosphate, adjusted to pH 3.0 with concentrated phosphoric acid) (58:42, v/v). The flow rate was set at 1.0 mL/min and the detection wavelength was set at 241 nm. The method was linear from 0.15 - 15.0 mg/L for both enantiomers. The limit of detection and the limit of quantification were 0.05 mg/L and 0.15 mg/L, respectively. The relative standard deviations (RSDs) of inter- and intra-day determination were below 0.5%. The method is easy to handle, accurate, and suitable for the quality control of the optical purity of alpha-phenylethylamine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamin, 98%
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamin, 98%
Sigma-Aldrich
α-Methylbenzylamin, 99%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamin, ChiPros®, produced by BASF, ≥99.0%
Supelco
(R)-(+)-α-Methylbenzylamin, derivatization grade (chiral), LiChropur, ≥99.0%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamin, purum, ≥98.0% (sum of enantiomers, GC)