Merck
Contact urticaria due to phenoxyethanol in an aftershave.
Dermatitis : contact, atopic, occupational, drug (2009-10-07)
Dunia Lujan, Buenaventura Hernandez-Machin, Yeray Peñate, Leopoldo Borrego
ZUSAMMENFASSUNG
The occurrence of pruritus immediately after application of an aftershave product is usually due to irritant contact dermatitis. We report a case of contact urticaria in a male patient, produced by an aftershave product containing phenoxyethanol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
2-Phenoxy-ethanol, ≥90%