Merck
Contact dermatitis from acrylics in a histology laboratory assistant.
Dermatitis : contact, atopic, occupational, drug (2009-10-08)
Luciana Molina, Antoine Amado, Peter L Mattei, James S Taylor
ZUSAMMENFASSUNG
The use of acrylics has expanded enormously, resulting in a vast range of products for both occupational and non-occupational purposes. Acrylics reported to cause allergic contact dermatitis in histology technicians are 2-hydroxyethyl methacrylate and 2-hydroxyethel acrylate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxyethylacrylat, 96%, contains 200-650 ppm monomethyl ether hydroquinone as inhibitor