- Efficient method for the synthesis of functionalized pyrazoles by catalyst-free one-pot tandem reaction of nitroalkenes with ethyl diazoacetate.
Efficient method for the synthesis of functionalized pyrazoles by catalyst-free one-pot tandem reaction of nitroalkenes with ethyl diazoacetate.
Organic & biomolecular chemistry (2009-10-16)
Jian-Wu Xie, Zheng Wang, Wei-Jun Yang, Li-Chun Kong, Dong-Cheng Xu
PMID19830281
ZUSAMMENFASSUNG
The one-pot synthesis of multisubstituted pyrazole derivatives was achieved via catalyst-free 1,3-dipolar cycloaddition of ethyl diazoacetate and nitroalkenes as the key step and elimination of the leaving group (NO(2) or Br) followed by intramolecular proton transfer with satisfactory yields.