Merck
HS-SPME-GC-MS analysis of body odor to test the efficacy of foot deodorant formulations.
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI) (2009-10-17)
Alessandro Caroprese, Simone Gabbanini, Claudia Beltramini, Elena Lucchi, Luca Valgimigli
ZUSAMMENFASSUNG
Foot malodor is mostly due to short-chain fatty acids produced by bacterial metabolism of eccrine sweating. We aimed to develop a protocol for an objective (instrumental) efficacy evaluation of foot deodorant formulations. Head-space solid-phase microextractions of target fatty acids from the feet of six healthy volunteers were analyzed by GC-MS. A comparative analysis of the treated vs. the untreated foot was performed in each subject after washing the feet with a physiologic solution and incubating at 36 degrees C for 24-72 h in tryptic soy agar growth medium. Acetic, butyric, isobutyric and isovaleric acids were identified as the main contributors to foot malodor in the majority of volunteers. Propionic, valeric and isocaproic acids were also detected in some subjects. Comparative analysis according to the protocol developed showed a statistically significant (P<0.01) reduction of target fatty acids ranging from -26.6% to -77.0%. The protocol developed is a convenient, sensitive and non-invasive method to test the efficacy of foot deodorant formulations in human volunteers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isovaleriansäure, 99%
Sigma-Aldrich
Isobuttersäure, 99%
Sigma-Aldrich
Isobuttersäure, ≥99%, FCC, FG
Supelco
Isobuttersäure, analytical standard
Sigma-Aldrich
Isovaleriansäure, ≥99%, FCC, FG
Sigma-Aldrich
Isovaleriansäure, natural, ≥98%, FG
Sigma-Aldrich
Isobuttersäure, natural, ≥99%, FCC, FG
Sigma-Aldrich
Isobuttersäure, puriss. p.a., ≥99.5%