Merck
Hypnotic and sleep quality-enhancing properties of kavain in sleep-disturbed rats.
Journal of pharmacological sciences (2009-11-03)
Ryuki Tsutsui, Kazuaki Shinomiya, Yasuhiro Takeda, Yoshihito Obara, Yoshihisa Kitamura, Chiaki Kamei
ZUSAMMENFASSUNG
The present study was performed to investigate the effects of kavain on the sleep-wake cycle in comparison with that of rilmazafone and diphenhydramine using sleep-disturbed rats. Electrodes for the electroencephalogram (EEG) and electromyogram (EMG) were implanted into Wistar rats. Total awake time, non-rapid eye movement (non-REM) sleep and rapid eye movement (REM) sleep were measured for 6 h. Kavain and rilmazafone showed a significant shortening in sleep latency, decreased awake time, and increased non-REM sleep time. On the other hand, significant shortening of the sleep latency was observed following the administration of diphenhydramine, while no effects were observed on the awake and non-REM sleep time. Moreover, kavain showed a significant increase in delta activity during non-REM sleep in sleep-disturbed rats, whereas a significant decrease in delta power during non-REM sleep was observed with rilmazafone. These results clearly indicate that kavain is a compound with not only hypnotic effects, but also sleep quality-enhancement effects.