Merck
  • Effect of oxidoreduction potential and of gas bubbling on rheological properties and microstructure of acid skim milk gels acidified with glucono-delta-lactone.
Effect of oxidoreduction potential and of gas bubbling on rheological properties and microstructure of acid skim milk gels acidified with glucono-delta-lactone.
Journal of dairy science (2009-11-20)
F Martin, N Cayot, A Marin, L Journaux, P Cayot, P Gervais, R Cachon
ZUSAMMENFASSUNG
Milk oxidoreduction potential was modified using gases during the production of a model dairy product and its effect on gel setting was studied. Acidification by glucono-delta-lactone was used to examine the physicochemistry of gelation and to avoid variations due to microorganisms sensitive to oxidoreduction potential. Four conditions of oxidoreduction potential were applied to milk: milk was gassed with air, nongassed, gassed with N(2), or gassed with N(2)H(2). The rheological properties and microstructure of these gels were determined using viscoelasticimetry, measurement of whey separation, and confocal laser scanning microscopy. It appeared that a reducing environment led to less-aggregated proteins within the matrix and consequently decreased whey separation significantly. The use of gas to modify oxidoreduction potential is a possible way to improve the quality of dairy products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Gluconsäure δ-Lacton, ≥99.0% (GC)
Sigma-Aldrich
Gluconolacton, 99.0-101.0%, meets USP testing specifications