Merck
Stabilizing labile DNA-protein complexes in polyacrylamide gels.
Electrophoresis (2010-01-29)
Nina Y Sidorova, Stevephen Hung, Donald C Rau
ZUSAMMENFASSUNG
The electrophoretic mobility-shift assay (EMSA) is one of the most popular tools in molecular biology for measuring DNA-protein interactions. EMSA, as standardly practiced today, works well for complexes with association binding constants K(a)>10(9) M(-1) under normal conditions of salt and pH. Many DNA-protein complexes are not stable enough so that they dissociate while moving through the gel matrix giving smeared bands that are difficult to quantitate reliably. In this work we demonstrate that the addition of the osmolyte triethylene glycol to polyacrylamide gels dramatically stabilizes labile restriction endonuclease EcoRI complexes with nonspecific DNA sequences enabling quantitation of binding using EMSA. The significant improvement of the technique resulting from the addition of osmolytes to the gel matrix greatly extends the range of binding constants of protein-DNA complexes that can be investigated using this widely used assay. Extension of this approach to other techniques used for separating bound and free components such as gel chromatography and CE is straightforward.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triethylenglycol, ReagentPlus®, 99%
Sigma-Aldrich
Triethylenglycol, BioUltra, anhydrous, ≥99.0% (GC)