Merck
  • Indoleamine 2,3-dioxygenase-expressing mature human monocyte-derived dendritic cells expand potent autologous regulatory T cells: consideration of triamterene to treat lupus.
Indoleamine 2,3-dioxygenase-expressing mature human monocyte-derived dendritic cells expand potent autologous regulatory T cells: consideration of triamterene to treat lupus.
Medical hypotheses (2010-02-02)
Richard E Kast, Eric L Altschuler
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triamteren, ≥99%