Merck
  • Induction of apoptosis by A3 adenosine receptor agonist N-(3-iodobenzyl)-adenosine-5'-N-methylcarboxamide in human leukaemia cells: a possible involvement of intracellular mechanism.
Induction of apoptosis by A3 adenosine receptor agonist N-(3-iodobenzyl)-adenosine-5'-N-methylcarboxamide in human leukaemia cells: a possible involvement of intracellular mechanism.
Acta physiologica (Oxford, England) (2010-02-04)
P Mlejnek, P Dolezel
ZUSAMMENFASSUNG
The sensitivity of cancer cells which exhibit multi-drug resistance phenotype to A3 adenosine receptor (A3AR) agonist N(6)-(3-iodobenzyl)-adenosine-5'-N-methylcarboxamide (IB-MECA) was studied. To establish direct relationship between P-glycoprotein (P-gp, ABCB1 and MDR1) expression and IB-MECA induced cell death, a straightforward method for precise estimation of intracellular level of this A3AR agonist was developed. We subjected three human leukaemia cell lines HL-60, K562 and K562/HHT to treatment with micromolar concentrations of IB-MECA. Although all cell lines used expressed A3AR, there was a large difference in their sensitivity to IB-MECA. While HL-60 and K562 cells were almost equally sensitive, the K562/HHT cells, which exhibit a multi-drug resistance phenotype because of overexpression of P-gp, were significantly more resistant. We found that the intracellular level of IB-MECA in K562/HHT cells was approx. 10 times lower than those in HL-60 or K562 cells. Inhibitors of P-gp, including cyclosporine A (CsA) and verapamil (Vpa), increased the intracellular level of IB-MECA and reversed the resistance of K562/HHT cells to this drug. Accordingly, shRNA-mediated down-regulation of P-gp significantly increased the intracellular level of IB-MECA in K562/HHT cells which simultaneously exhibited reduced resistance to this A3AR agonist. In addition, an in vitro enzyme-based assay provided evidence that IB-MECA might serve as a substrate for P-gp. Our results suggest that P-gp overexpression prevents cells from IB-MECA induced apoptosis despite the A3AR expression. Pro-apoptotic effect of IB-MECA seemed to strongly depend on its intracellular accumulation rather than on its interaction with A3AR.