Merck
  • Antigenic changes in human albumin caused by reactivity with the occupational allergen diphenylmethane diisocyanate.
Antigenic changes in human albumin caused by reactivity with the occupational allergen diphenylmethane diisocyanate.
Analytical biochemistry (2010-02-04)
Adam V Wisnewski, Jian Liu, Carrie A Redlich
ZUSAMMENFASSUNG
Diphenylmethane diisocyanate (MDI), the chemical commonly used as a cross-linking agent in commercial polyurethane production, is a well-recognized cause of asthma. Reaction products between MDI and "self" proteins are hypothesized to act as antigens capable of inducing airway inflammation and asthma; however, such MDI antigens remain incompletely understood. We used a variety of analytical methods to characterize the range of MDI-albumin reaction products that form under physiological conditions. Sites of MDI conjugation on antigenic MDI-albumin products, as defined by serum immunoglobulin G (IgG) from MDI-exposed workers, were determined by high-performance liquid chromatography (HPLC) followed by tandem mass spectrometry (MS/MS). The data identified 14 MDI conjugation sites (12 lysines and 2 asparagines) on human albumin and highlight reaction specificity for the second lysine in dilysine (KK) motifs, and this may be a common characteristic of "immune-sensitizing" chemicals. Several of the MDI conjugation sites are not conserved in albumin from other species, and this may suggest species differences in epitope specificity for self protein (albumin)-isocyanate conjugates. The study also describes new applications of contemporary proteomic methodology for characterizing and standardizing MDI-albumin conjugates destined for use in clinical research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,4′-Methylen-bis(phenylisocyanat), 98%
Supelco
4,4′-MDI, analytical standard