- Use of polystyrene-supported 2-isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline for the preparation of a hapten-protein conjugate for antibody development.
Use of polystyrene-supported 2-isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline for the preparation of a hapten-protein conjugate for antibody development.
Bioorganic & medicinal chemistry letters (2010-02-05)
Manisha Sathe, Mariliza Derveni, Marjorie Allen, David C Cullen
PMID20129781
ZUSAMMENFASSUNG
Polystyrene-supported 2-isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline (PS-IIDQ), a polymer-supported covalent coupling reagent, was successfully employed for the first time in the bioconjugation of an example hapten (phytanic acid derivative) to a carrier protein (bovine serum albumin (BSA)) within the context of immunogen preparation for antibody development. The ability of the prepared example phytanic acid derivative-BSA conjugate to bind an anti-phytanic acid antibody was confirmed using an enzyme-linked immunosorbent assay (ELISA).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.