Merck
  • Total synthesis of (+)-chaetocin and its analogues: their histone methyltransferase G9a inhibitory activity.
Total synthesis of (+)-chaetocin and its analogues: their histone methyltransferase G9a inhibitory activity.
Journal of the American Chemical Society (2010-03-10)
Eriko Iwasa, Yoshitaka Hamashima, Shinya Fujishiro, Eisuke Higuchi, Akihiro Ito, Minoru Yoshida, Mikiko Sodeoka
ZUSAMMENFASSUNG
The first total synthesis of (+)-chaetocin has been accomplished in nine steps starting from known N-Cbz-N-Me-serine using radical alpha-bromination reaction of diketopiperazine 10 and Co(I)-mediated reductive dimerization reaction of 12 as key reactions. The enantiomers show comparable inhibitory activity toward histone methyltransferase (HMT) G9a, but analogues without the sulfur functionality are inactive.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chaetocin aus Chaetomium minutum, ≥95% (HPLC)