Merck
  • Pressure study on symmetric and asymmetric phospholipid bilayers: effect of vesicle size on Prodan fluorescence.
Pressure study on symmetric and asymmetric phospholipid bilayers: effect of vesicle size on Prodan fluorescence.
Annals of the New York Academy of Sciences (2010-03-18)
Masaki Goto, Masataka Kusube, Makoto Nishimoto, Nobutake Tamai, Hitoshi Matsuki, Shoji Kaneshina
ZUSAMMENFASSUNG
The bilayer phase behavior of symmetric and asymmetric phosphatidylcholines (PCs), 1,2-diheptadecanoyl-PC (C17PC), 1-palmitoyl-2-stearoyl-PC (PSPC), and 1-stearoyl-2-palmitoyl-PC (SPPC), with different vesicle sizes were investigated by a high-pressure fluorescence method using the polarity-sensitive fluorescent probe Prodan. The second derivative of fluorescence spectra for all the PCs of small-sized vesicle showed four minima characteristic of four membrane states on the spectra irrespective of the acyl-chain symmetry, whereas those of large-sized vesicle had one more minimum originating from the most hydrophilic site at the membrane surface. These findings indicate that Prodan molecules can distribute into multiple sites in the bilayer and move around the head-group region depending on the vesicle size. The behavior of the spectra in the SPPC bilayer suggested that the interdigitated gel phase had a less polar "pocket" formed by a space between uneven terminal methyl ends of the sn-1 and sn-2 chains. It turned out that the curvature of vesicles affects the distribution of the Prodan molecules in all phases, more particularly in the interdigitated gel phase.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethyl-6-propionyl-2-naphthylamin, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)