Merck
Diarrhetic shellfish toxins: improvement of sample clean-up for HPLC determination.
Toxicon : official journal of the International Society on Toxinology (1991-01-01)
O B Stabell, V Hormazabal, I Steffenak, K Pedersen
ZUSAMMENFASSUNG
Okadaic acid and dinophysistoxin-1, the principal toxic components in diarrhetic shellfish poisoning, may be detected by high-performance liquid chromatography and fluorometric measurement as 9-anthrylmethyl esters. However, "greasy" samples may occur and the fluorescent reagent 9-anthryldiazomethane may decompose during storage, resulting in impurities that may seriously interfere with quantitative determination. Ultrasonic treatment of the samples during derivatization with 9-anthryldiazomethane was found to improve reproducibility. This may result from increased access to reactive sites on toxins by 9-anthryldiazomethane due to disruption of micelles formed by toxins and other partly hydrophobic compounds. A procedure for cleaning the derivatized samples, using a 0.1 g silica cartridge column and different eluent compositions from that reported by LEE et al. (1987), was found to facilitate chromatogram interpretation. Deoxycholic acid, a commercial available bile acid, was found to be an acceptable internal standard. The 9-anthrylmethyl esters of okadaic acid, dinophysistoxin-1 and deoxycholic acid, were stable at 4 degrees C for at least seven days when stored dry or in methanol.