- Polyneuropathy induced by m-tolyl methyl carbamate intoxication.
Polyneuropathy induced by m-tolyl methyl carbamate intoxication.
Journal of neurology (1991-02-01)
F Umehara, S Izumo, K Arimura, M Osame
PMID2030373
ZUSAMMENFASSUNG
A 55-year-old woman who attempted suicide by ingesting 200 ml of m-tolyl methyl carbamate (MTMC) is reported. She was comatose for 3 days and, upon recovery, had notable paraesthesia in her lower limbs and difficulty in walking. Neurological examination revealed sensorimotor polyneuropathy. Sural nerve biopsy revealed marked axonal degeneration with a moderate decrease of myelinated fibres.