Merck
  • The structure of oleamide films at the aluminum/oil interface and aluminum/air interface studied by Sum Frequency Generation (SFG) vibrational spectroscopy and Reflection Absorption Infrared Spectroscopy (RAIRS).
The structure of oleamide films at the aluminum/oil interface and aluminum/air interface studied by Sum Frequency Generation (SFG) vibrational spectroscopy and Reflection Absorption Infrared Spectroscopy (RAIRS).
ACS applied materials & interfaces (2010-04-02)
Michael T L Casford, Paul B Davies
ZUSAMMENFASSUNG
The structure of oleamide (cis-9-octadecenamide) films on aluminum has been investigated by sum frequency generation vibrational spectroscopy (SFG) and reflection absorption infrared spectroscopy (RAIRS). Three different film deposition strategies were investigated: (i) films formed by equilibrium adsorption from oleamide solutions in oil, (ii) Langmuir-Blodgett films cast at 1 and 25 mN m(-1), (iii) thick spin-cast films. Both L-B and spin-cast films were examined in air and under oil. The adsorbate formed in the 1 mN m(-1) film in air showed little orientational order. For this film, the spectroscopic results and the ellipsometric thickness point to a relatively conformationally disordered monolayer that is oriented principally in the plane of the interface. Direct adsorption to the metal interface from oil results in SFG spectra of oleamide that are comparable to those observed for the 1 mN m(-1) L-B film in air. In contrast, SFG and RAIRS results for the 25 mN m(-1) film in air and SFG spectra of the spin-cast film in air both show strong conformational ordering and orientational alignment normal to the interface. The 25 mN m(-1) film has an ellipsometric thickness almost twice that of the 1 mN m(-1) L-B film. Taken in combination with the spectroscopic results, this is indicative of a well packed monolayer in air in which the hydrocarbon chain is in an essentially defect-free extended conformation with the methyl terminus oriented away from the surface. A similar structure is also deduced for the surface of the spin-cast film in air. Upon immersion of the 25 mN m(-1) L-B film in oil the SFG spectra show that this film rapidly adopts a relatively disordered structure similar to that seen for the 1 mN m(-1) L-B film in air. Immersion of the spin-cast film in oil results in the gradual disordering of the amide film over a period of several days until the observed spectra become essentially identical to those observed for direct adsorption of oleamide from oil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ölsäureamid, ≥99%