Merck
  • An ultra-high-performance liquid chromatography-tandem mass spectrometry method for simultaneous determination of aflatoxins B1, B2, G1, G2, M1 and M2 in traditional Chinese medicines.
An ultra-high-performance liquid chromatography-tandem mass spectrometry method for simultaneous determination of aflatoxins B1, B2, G1, G2, M1 and M2 in traditional Chinese medicines.
Analytica chimica acta (2010-04-07)
Zheng Han, Yunliang Zheng, Lianjun Luan, Zengxuan Cai, Yiping Ren, Yongjiang Wu
ZUSAMMENFASSUNG
An ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) method for simultaneous determination of aflatoxins B1, B2, G1, G2, M1 and M2 in traditional Chinese medicines (TCMs) was developed. The approach was characterized in details and a special focus was placed on the recovery rates of isolation procedure in different TCM matrices, i.e. rhizomes and roots, seeds, flowers, grasses and leaves. For this purpose, [(13)C(17)]-aflatoxinB1 was employed as the internal standard and a reliable solid phase extraction-based clean-up method was developed. The observed recovery rates of the six aflatoxins ranged from 85.6% to 117.6% in different matrices. Then, the established method was successfully applied to the determination of the six aflatoxins in various TCMs. For 30 commercial samples analyzed, 16 were contaminated with aflatoxins. The mean levels (incidence) of aflatoxins B1, B2, G1 and G2 in positive samples were 1.40 (68.8%), 1.27 (50.0%), 0.50 (43.8%) and 0.94 (43.8%) microg kg(-1), respectively. Interestingly, aflatoxin M1 was detected in two samples with the maximal content of 0.70 microg kg(-1). No sample was contaminated with aflatoxin M2. Meanwhile, a possible association between the contamination levels and the selected herbs was clarified in the present study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aflatoxin G2
Sigma-Aldrich
Aflatoxin B2
Sigma-Aldrich
Aflatoxin G1 aus Aspergillus flavus, from Aspergillus flavus
Supelco
Aflatoxin B2 -Lösung, 0.5 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin G2 -Lösung, 0.5 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin G1 -Lösung, 2 μg/mL in acetonitrile, analytical standard
Aflatoxin G2 -Lösung, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin B2 -Lösung, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin G1 -Lösung, 3.78 μg/g in acetonitrile, ERM®, certified reference material