Merck
  • Identification and structure of small-molecule stabilizers of 14-3-3 protein-protein interactions.
Identification and structure of small-molecule stabilizers of 14-3-3 protein-protein interactions.
Angewandte Chemie (International ed. in English) (2010-05-04)
Rolf Rose, Silke Erdmann, Stefanie Bovens, Alexander Wolf, Micheline Rose, Sven Hennig, Herbert Waldmann, Christian Ottmann
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fusicoccin from Fusicoccum amygdali, ≥85% (HPLC)