Merck
Evaluation of [(11)C]metergoline as a PET radiotracer for 5HTR in nonhuman primates.
Bioorganic & medicinal chemistry (2010-05-11)
Jacob M Hooker, Sung Won Kim, Achim T Reibel, David Alexoff, Youwen Xu, Colleen Shea
ZUSAMMENFASSUNG
Metergoline, a serotonin receptor antagonist, was labeled with carbon-11 in order to evaluate its pharmacokinetics and distribution in non-human primates using positron emission tomography. [(11)C]Metergoline had moderate brain uptake and exhibited heterogeneous specific binding, which was blocked by pretreatment with metergoline and altanserin throughout the cortex. Non-specific binding and insensitivity to changes in synaptic serotonin limit its potential as a PET radiotracer. However, the characterization of [(11)C]metergoline pharmacokinetics and binding in the brain and peripheral organs using PET improves our understanding of metergoline drug pharmacology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Metergolin