Merck
  • Apoptosis-inducing action of two products from oxidation of sesamol, an antioxidative constituent of sesame oil: a possible cytotoxicity of oxidized antioxidant.
Apoptosis-inducing action of two products from oxidation of sesamol, an antioxidative constituent of sesame oil: a possible cytotoxicity of oxidized antioxidant.
Toxicology in vitro : an international journal published in association with BIBRA (2010-06-01)
Aya Fujimoto, Yoshimi Shingai, Toshihisa B Oyama, Takuya Kawanai, Erika Hashimoto, Kazuki Koizumi, Kaori Kimura, Toshiya Masuda, Yasuo Oyama
ZUSAMMENFASSUNG
Many effects of sesamol, an antioxidative constituent of sesame oil, have been reported for human health benefits due to its antioxidative action. However, we recently isolated two cytotoxic products, trimer and tetramer of sesamol, from oxidation of sesamol by an assay-guided purification. In this study, we have revealed some cytotoxic characteristics of these products in rat thymocytes and human leukemia K562 cells. Incubation of cells with trimer or tetramer at 10-30 microM for 24h significantly increased cell lethality and population of rat thymocytes containing hypodiploid DNA, suggesting cell death with DNA fragmentation, while it was not the case for 30 microM sesamol. The cytotoxic action of tetramer was more potent than that of trimer in rat thymocytes when their concentrations were 10-30 microM. The incubation of cells with 10 microM tetramer for 24h increased the population of cells with exposed phosphatidylserine, the activity of caspases, and the nick of DNA. These results indicate tetramer-induced apoptosis. In K562 cells, the incubation with tetramer at 10 microM for 72 h significantly inhibited the growth without affecting the lethality. However, tetramer at 30 microM significantly increased cell lethality. It is likely that tetramer exerts more cytotoxic action on normal non-proliferative cells (rat thymocytes) rather than proliferative cancer cells (human leukemia K562 cells). It may be necessary to consider the condition for preservation of sesamol and the safety of products from in vivo oxidation of sesamol for human health.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sesamol, 98%