Merck
Kyotorphin transport and metabolism in rat and mouse neonatal astrocytes.
Brain research (2010-06-12)
Jianming Xiang, Huidi Jiang, Yongjun Hu, David E Smith, Richard F Keep
ZUSAMMENFASSUNG
Neuropeptide inactivation is generally thought to occur via peptidase-mediated degradation. However, a recent study found increased analgesia after L-kyotorphin (L-Tyr-L-Arg; L-KTP) administration in mice lacking an oligopeptide transporter, PEPT2. The current study examines the role of PEPT2 in L-KTP uptake by astrocytes and compares it to astrocytic L-KTP degradation. L-[(3)H]KTP uptake was measured in primary cultures of neonatal astrocytes from rats and from Pept2(+/+) and Pept2(-/-) mice. Uptake was further characterized using potential inhibitors. L-[(3)H]KTP degradation was examined in primary astrocyte cultures from Pept2(-/-) mice by following the formation of L-[(3)H]tyrosine. The uptake of L-[(3)H]KTP in both rat and Pept2(+/+) mouse neonatal astrocytes was inhibited by known PEPT2 inhibitors. L-[(3)H]KTP uptake was also reduced in Pept2(-/-) astrocytes as compared to those from Pept2(+/+) mice. Kinetic analysis indicated the presence of a high affinity (K(m) approximately 50 microM) transporter for L-[(3)H]KTP, identified as Pept2, and a low affinity transporter (K(m) approximately 3-4 mM), inhibited by amastatin, bestatin and tyrosine. Astrocytes also degraded L-KTP through a low affinity peptidase (K(m) approximately 2 mM). Astrocytic clearance of L-KTP occurs via both peptidase activity and transport. These processes occur at similar rates and may be linked. This supports the contention that oligopeptide transport may have an impact on the extracellular clearance (and potentially activity) of certain neuropeptides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Cefadroxil, analytical standard