Merck
  • Analysis of carbohydrate and fatty acid marker abundance in ricin toxin preparations for forensic information.
Analysis of carbohydrate and fatty acid marker abundance in ricin toxin preparations for forensic information.
Analytical chemistry (2010-06-24)
Heather A Colburn, David S Wunschel, Helen W Kreuzer, James J Moran, Kathryn C Antolick, Angela M Melville
ZUSAMMENFASSUNG
One challenge in the forensic analysis of ricin samples is determining the method and extent of sample preparation. Ricin purification from the source castor seeds is essentially a protein purification through removal of the nonprotein fractions of the seed. Two major, nonprotein constituents in the seed are the castor oil and carbohydrates. We used derivatization of carbohydrate and fatty acid markers followed by identification and quantification using gas chromatography/mass spectrometry (GC/MS) to assess compositional changes in ricin samples purified by different methods. The loss of ricinoleic acid indicated steps for oil removal had occurred, and a large decrease of ricinoleic acid was observed between unextracted mash and solvent extracted and protein precipitate preparations. Changes to the carbohydrate content of the sample were also observed following protein precipitation. The differential loss of arabinose relative to mannose was observed indicating the removal of the major carbohydrate fraction of the seed and enrichment of the protein content. When the data is combined and multivariate principle component analysis is applied, these changes in fatty acid and carbohydrate abundance are discriminating enough to be indicative of the preparation method used for each sample.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rizinolsäure, ≥95%