- Discovery process and characterization of novel carbohydrazide derivatives as potent and selective GHSR1a antagonists.
Discovery process and characterization of novel carbohydrazide derivatives as potent and selective GHSR1a antagonists.
ChemMedChem (2010-07-02)
Fabio Maria Sabbatini, Romano Di Fabio, Mauro Corsi, Paolo Cavanni, Steve M Bromidge, Yves St-Denis, Lucilla D'Adamo, Stefania Contini, Marilisa Rinaldi, Sebastien Guery, Chiara Savoia, Claudia Mundi, Benedetta Perini, Andrew J Carpenter, Giovanna Dal Forno, Federico Faggioni, Michela Tessari, Francesca Pavone, Carla Di Francesco, Alberto Buson, Mario Mattioli, Elisabetta Perdona', Sergio Melotto
PMID20593439
MATERIALIEN