Merck
[The treatment of MRSA colonized middle ear; case report and literature review].
Laryngo- rhino- otologie (2010-07-10)
G Senska, D Rüsing
ZUSAMMENFASSUNG
The treatment of MRSA (methocillin resistant staphylococcus aureus) colonized middle ear is difficult. According to the guidelines, a MRSA colonized Patient is not to be treated with systemic antibiotics. The topical treatment shows the problem of the ototoxicity of most of the used antiseptic as well as antibiotic substances. Selective literature review and consideration of the author's own clinical experience. Antibiotic treatment options include aequeous Tetracyclin drops, aequeous chloramphenicol drops and quinolon ear drops (unfortunately the MRSA is resistent mostly). Antiseptics without ototoxic effects are the Burow's solution, Povidone-iode, acetic acid solutions and aequeous dequalinium solutions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Dequaliniumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material