Merck
  • Convenient and efficient microwave-assisted synthesis of a methyl derivative of the fused indoloquinoline alkaloid cryptosanguinolentine.
Convenient and efficient microwave-assisted synthesis of a methyl derivative of the fused indoloquinoline alkaloid cryptosanguinolentine.
Molecules (Basel, Switzerland) (2010-07-27)
Robert M Gengan, Pitchai Pandian, Chandraprakash Kumarsamy, Palathurai S Mohan
ZUSAMMENFASSUNG
An efficient synthesis of a methyl derivative of the indoloquinoline alkaloid cryptosanguinolentine based on microwave-assisted reactions is described. The microwave-assisted synthesis of an intermediate 4-hydroxy-2-methylquinoline yielded 86% of the desired product and other intermediates prepared yielded high % of products in shorter reaction times, under optimum conditions, as compared to traditional methods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chinaldin, ≥95.0% (GC)
Supelco
Chinaldin, analytical standard, ≥90% (GC)