Merck
  • The acid tolerant L-arabinose isomerase from the food grade Lactobacillus sakei 23K is an attractive D-tagatose producer.
The acid tolerant L-arabinose isomerase from the food grade Lactobacillus sakei 23K is an attractive D-tagatose producer.
Bioresource technology (2010-08-07)
Moez Rhimi, Rimeh Ilhammami, Goran Bajic, Samira Boudebbouze, Emmanuelle Maguin, Richard Haser, Nushin Aghajari
ZUSAMMENFASSUNG
The araA gene encoding an L-arabinose isomerase (L-AI) from the psychrotrophic and food grade Lactobacillus sakei 23K was cloned, sequenced and over-expressed in Escherichia coli. The recombinant enzyme has an apparent molecular weight of nearly 220 kDa, suggesting it is a tetramer of four 54 kDa monomers. The enzyme is distinguishable from previously reported L-AIs by its high activity and stability at temperatures from 4 to 40 degrees C, and pH from 3 to 8, and by its low metal requirement of only 0.8 mM Mn(2+) and 0.8 mM Mg(2+) for its maximal activity and thermostability. Enzyme kinetic studies showed that this enzyme displays a high catalytic efficiency allowing D-galactose bioconversion rates of 20% and 36% at 10 and 45 degrees C, respectively, which are useful for commercial production of D-tagatose.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(−)-Tagatose, ≥98% (HPLC)