Merck
'Black Salve' and melanomas.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS (2010-08-14)
C E Payne
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Antimon(III)-sulfid, 99.995% trace metals basis
Sigma-Aldrich
Antimon(III)-sulfid, powder