Merck
  • Efficient production and purification of functional bacteriorhodopsin with a wheat-germ cell-free system and a combination of Fos-choline and CHAPS detergents.
Efficient production and purification of functional bacteriorhodopsin with a wheat-germ cell-free system and a combination of Fos-choline and CHAPS detergents.
Biochemical and biophysical research communications (2010-09-03)
Takahisa Genji, Akira Nozawa, Yuzuru Tozawa
ZUSAMMENFASSUNG
Cell-free translation is one potential approach to the production of functional transmembrane proteins. We have now examined various detergents as supplements to a wheat-germ cell-free system in order to optimize the production and subsequent purification of a functional model transmembrane protein, bacteriorhodopsin. We found that Fos-choline and CHAPS detergents counteracted each other's inhibitory effects on cell-free translation activity and thereby allowed the efficient production and subsequent purification of functional bacteriorhodopsin in high yield.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CHAPS 100 mM -Lösung